Blender Base for Waring* Blenders
Catalog #: 58983-050
Supplier: Eberbach
Customers Who Bought This Also Bought
Hydrogen peroxide 30% stabilized ACS, VWR Chemicals BDH®
Supplier: VWR International
$440.62 to $1,762.55
Bromophenol blue 0.04% (w/v) in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
$91.88 to $955.56
Conductivity Standards, Potassium Chloride (KCl) Solutions, Thermo Scientific
Supplier: Thermo Scientific Chemicals
$222.01