Temperature Gauge
Catalog #: 58700-322
Supplier: Elmer Schultz Services
Customers Who Bought This Also Bought
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$635.20 to $2,541.50
Formalin 10%, VWR®, Reagent Grade for histology, neutral buffered
Supplier: VWR International
$109.33 to $364.87
Accessories for Advanced® Multi-Sample Osmometer, Model 2020, Advanced Instruments
Supplier: Advanced Instruments
$2,423.69
Stablcal® Turbidity Standards Calibration Kit, 2100Q Portable Turbidimeter, Hach
Supplier: Hach
$949.82
o-Xylene reference substance for gas chromatography, Supelco®
Supplier: MilliporeSigma
$657.25 to $149,474.13