ALUMINA GF 10X20CM 250UM PK25
Catalog #: 57457-160
Supplier: Analtech
Customers Who Bought This Also Bought
Glycerine ≥99.7%, Laboratory Reagent, VWR Chemicals BDH®
Supplier: VWR International
$881.80 to $2,807.56
Sodium hydroxide 1.0 N in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
$72.65 to $846.50
Potassium dihydrogen phosphate ≥99.0% ACS, VWR Chemicals BDH®
Supplier: VWR International
$360.25 to $1,246.54