Order Entry
ContactUsLinkComponent
Full Height Sieve, Stainless Steel Wire and Frame
Full Height Sieves, Stainless Steel Wire and Frame, W.S. Tyler™
Catalog #: 57324-443
Supplier:  W.S. Tyler
Full Height Sieves, Stainless Steel Wire and Frame, W.S. Tyler™
Catalog #: 57324-443
Supplier:  W.S. Tyler

Specifications

  • Mesh Size:
    106 µm (0.0042")
  • USA Series:
    #140
  • Supplier No.:
    5211

Specifications

About this item

Full height sieves are 5.08cm (2") high above stainless steel wire cloth

Each sieve is serialized and supplied with a “Test Sieve Certificate.” Sieves are marked with the USA Sieve Number. Sieves one millimeter and coarser are identified by opening in millimeters and inches; sieves finer, by opening in microns and inches.

Conform to ASTM E-11 specifications.

Overall height: 7.4cm (2.9"). Diameter: 20.3cm (8").

Customers Who Bought This Also Bought