Lab Guard® Reclosable Biohazard Bags
Catalog #: 56766-372
Supplier: Minigrip
Specifications
- W×L:203×254 mm (8×10")
- Description:Biohazard bag
- Cat. no.:56766-372
- Packaging:500/dispenser box
Specifications
About this item
These specimen handling bags protect both workers and paperwork.
- Re-closable zipper for secondary leakage containment
- Additional marking areas identify contents needing to be kept frozen, refrigerated, or at room temperature
Many Lab Guard® bags now feature a double zipper for added closure security. Bags include document pouch.
They feature bright orange and black graphics to clearly identify bags as containing biohazardous materials. New destroyable biohazard symbol bags won't leak. They tear open easily at processing and can be disposed of in regular waste for significant cost savings.
The 3-wall bags contain an outside panel for storing and protecting paperwork. The 4-wall bags provide an additional wall of protection between paperwork and the contents of the bag.
Ordering information: For additional sizes, please contact your Avantor sales representative.
Customers Who Bought This Also Bought