Chemical Danger Series Signs and Labels, National Marker
Catalog #: 56606-714
Supplier: ACCUFORM MANUFACTURING, INC
Specifications
- Material:Vinyl, pressure-sensitive
- Legend:Acid
- Dimensions:17.8×25.4 cm (7×10")
- Supplier No.:D5P
Specifications
About this item
Chemical danger series signs alert employees to potential chemical hazards in the workplace by identifying chemical storage areas
Signs are red, white, and black, and indicate the danger with bold black type under the word DANGER. Meet OSHA 29 CFR-1910.145 and ANSI Z35.1.
Customers Who Bought This Also Bought