ContactUsLinkComponent
PIPETTE TIPS FINNTIP 5ML CS270
Finntip™ Pipette Tips, Thermo Scientific
Catalog #: 53515-980
Finntip™ Pipette Tips, Thermo Scientific
Catalog #: 53515-980
Specifications
  • Volume:
    1000 - 5000 µL
  • Packed:
    5×54/Rack
  • Tip Style:
    Finntip™ 5 mL
  • Supplier No.:
    9402070
Specifications
About this item
These high-quality pipette tips are specifically designed to fit Finnpipette® pipettors, but are also compatible with most pipettor brands.
  • Made of high-quality polypropylene
  • Autoclavable
  • Chemical-resistant
Tips in refill trays can be used to quickly refill existing racks.
Customers Who Bought This Also Bought