Finntip Stepper Tips, Thermo Scientific
Catalog #: 53515-932
Supplier: Thermo Fisher Scientific
Specifications
- Volume:500 µl, 1000 µl, 1500 µl, 2000 µl, 2500 µl
- Tip Style:Finntip Stepper 25 mL
- Supplier No.:9404220
Specifications
About this item
Finntip stepper tips are the perfect fit with Thermo Scientific Finnpipette stepper pipetters.
The finntip stepper tips are plunger-style tips for positive displacement pipetting. Seven sizes are available in sterile and non sterile versions.
Customers Who Bought This Also Bought