Wiretrol® II Disposable Micropipettes, Drummond
Catalog #: 53507-430
Supplier: Drummond
Specifications
- Capacity:10/20 µl
- Color:Orange
- Description:Wiretrol® II Micropipet
- Cat. no.:53507-430
- Supplier No.:5-000-2020
Specifications
About this item
This economical positive-displacement micropipette features fingertip control.
- Dual graduations for non-contact delivery
- Ideal for a wide variety of applications including both high- and low-viscosity liquids and corrosives
- Economical
The Wiretrol II provides the accuracy and convenience of the Wiretrol I series with dual graduations and a shorter plunger for non-contact delivery.
Customers Who Bought This Also Bought