Dialamatic Microdispensers, Drummond
Catalog #: 53506-274
Supplier: Drummond
Specifications
- Volume:5 - 25 µl
- Description:Dialamatic microdispenser, adjustable
- Increments:1 µl
- Accuracy:±1%
- Supplier No.:3-000-225
Specifications
About this item
Dialamatic Microdispensers feature borosilicate glass bores with stainless steel plungers.
- Borosilicate glass bores with stainless steel plungers (PTFE tips on all plungers 25 µl or larger)
- Adjustable, direct reading plunger sets volume
- Delivery scale engraved on plunger shaft
- Fractional deliveries possible with pre-marked indexes
- Volumetric accuracy of ±1% at full volume
The microdispensers have a volumetric accuracy of ±1% at full volume.
The direct reading plunger sets the volume, and the plunger shaft includes delivery-scale engravings.
Customers Who Bought This Also Bought