Accessories and Replacement Parts for VWR® Microdispensers
Catalog #: 53498-162
Supplier: VWR International
Specifications
- Description:Replacement Fluoropolymer Resin-Coated Tips
- Cat. no.:53498-162
- For Use With:1000 µL microdispensers
Specifications
About this item
VWR® pipette tips and capillaries are for use with VWR® microdispensers (see 53506-100 series).
- For use with VWR® microdispensers
Glass capillaries are color-coded, except for the 1000 µl size, which is made of natural polypropylene. The 20 µl capillary can only be used with VWR® 20 µl microdispenser (53506-530).
Customers Who Bought This Also Bought