Order Entry
ContactUsLinkComponent
Diamond Scriber
Diamond Scriber
Catalog #: 52865-005
Diamond Scriber
Diamond Scriber
Catalog #: 52865-005

Specifications

  • Description:
    Diamond scriber
  • Cat. no.:
    52865-005
  • Length:
    152 mm (6")
  • Supplier No.:
    200-22A

Specifications

About this item

For permanently marking glass slides, test tubes, and laboratory glassware

Sharp, selected diamond mounted in a round, clear aluminum handle tapered to fit hand comfortably. Tip retracts for protection and safety.

Ordering information: Also available in blue, black, red, and silver handles; contact your Avantor sales representative for more information.
Customers Who Bought This Also Bought