Order Entry
ContactUsLinkComponent
PVC Film Wrap
PVC Film Wrap, T FRANK MCCALLS INC
Catalog #: 52857-903
Supplier:  TFrank McCalls
PVC Film Wrap
PVC Film Wrap, T FRANK MCCALLS INC
Catalog #: 52857-903
Supplier:  TFrank McCalls

Specifications

  • Length:
    914.4 m (3000')
  • Width:
    457 mm (18")
  • Description:
    PVC film wrap, clear
  • Cat. no.:
    52857-903
  • Supplier No.:
    44104SEL3000

Specifications

About this item

All-purpose, clear, polyvinyl wrap

Self-sealing. Adheres readily to glass vessels such as test tubes, flasks, cylinders, bottles, petri dishes, etc. Supplied in plain white carton.

Customers Who Bought This Also Bought