Order Entry
ContactUsLinkComponent
VETERINARY REFRACTOMETER
Rhino VET 360 Veterinary Refractometer, Reichert
Catalog #: 51100-100
VETERINARY REFRACTOMETER
Rhino VET 360 Veterinary Refractometer, Reichert
Catalog #: 51100-100

Specifications

  • Description:
    Rhino VET 360 Veterinary Refractometer
  • Height:
    38 mm
  • Model:
    VET 360
  • Scales:
    Urine Specific Gravity: Small Animals,
    Urine Specific Gravity: Large Animals,
    Serum Protein
  • Cat. no.:
    51100-100
  • Accuracy:
    Urine specific gravity: ±0.001, serum protein: ±0.2 g/100 ml
  • Length:
    235 mm
  • Range:
    Urine specific gravity: 1.000 - 1.060, serum protein: 2.0 - 14.0 g/100ml
  • Weight:
    0.16 kg
  • Width:
    38 mm
  • Supplier No.:
    137536L0

Specifications

About this item

The Rhino-rugged water resistant, dustproof, and shock resistant hand-held refractometer uses Automatic Temperature Compensation to provide accurate readings regardless of ambient temperature.

  • Automatically corrects all readings back to the Standard Reference Temperature of 68°F (20°C) providing greater accuracy
  • Improved optics that are easy to read
  • No batteries required making it reliable any time
  • Ideal for use in the office or outdoors
  • Large and small animal scales in one instrument
  • IP67 rated, sealed internal optics cannot be contaminated
  • Test takes only seconds
Customers Who Bought This Also Bought