Order Entry
ContactUsLinkComponent
Mortar and Pestle Set, Glass
Mortar and Pestle Sets, Glass, Variety Glass
Catalog #: 50415-020
Supplier:  Variety Glass
Mortar and Pestle Set, Glass
Mortar and Pestle Sets, Glass, Variety Glass
Catalog #: 50415-020
Supplier:  Variety Glass

Specifications

  • Volume:
    57 ml
  • Height:
    68 mm
  • Inner Diameter:
    80 mm
  • Supplier No.:
    826-2

Specifications

About this item

Mortar and pestle sets are made of clear, heavy, flint glass

Mortar has a heavy molded base and lip for pouring. Sold as a set.

Customers Who Bought This Also Bought