REGULATOR ANAL 1STAGE NDL CGA 350 100PSI
Catalog #: 47752-062
Supplier: Airgas
Customers Who Bought This Also Bought
Target™ All-Plastic Disposable Luer-Lok Syringes, Thermo Scientific
Supplier: Thermo Scientific
233,35 $ to 2 174,33 $
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
684,63 $ to 2 738,51 $
Sodium hydroxide solution 50% (w/w), BAKER ANALYZED®, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
231,20 $ to 2 774,16 $
L(+)-Ascorbic acid 99.0-100.5% USP, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
361,74 $ to 2 107,25 $