REGULATOR ANAL 1STAGE CGA 590 0 25 PSI
Catalog #: 47752-054
Supplier: Airgas
Customers Who Bought This Also Bought
Sodium hydroxide 50% (w/w) in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
684,53 $
Sodium hydroxide 0.02 N in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
73,05 $ to 828,17 $
MiniCollect* Tube K2E K2Edta Non sterile Lavender Cap 0.25/0.5 Ml PK500
Supplier: Greiner Bio-One
656,98 $ to 2 627,82 $
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
684,63 $ to 2 738,51 $
Choline chloride 98.0-100.5% (by anhydrous basis), BAKER ANALYZED®, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
149,28 $
4-(Methoxycarbonyl)phenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
174,73 $
2,2'-Azobis[2-(2-imidazolin-2-yl)propane] dihydrochloride ≥98.0% (by HPLC)
Supplier: TCI America
58,73 $