Order Entry
ContactUsLinkComponent
Sterileware* Red Scoop
SP Bel-Art Sterileware® Red Sterile Sampling Scoops, Bel-Art Products, a part of SP
Catalog #: 47751-008
Sterileware* Red Scoop
SP Bel-Art Sterileware® Red Sterile Sampling Scoops, Bel-Art Products, a part of SP
Catalog #: 47751-008

Specifications

  • Capacity:
    250 ml (8.5 oz.)
  • Bowl Dimensions (W×L×H):
    13.5×5.4×4.9 cm (5⁵⁄₁₆×2⅛×1¹⁵⁄₁₆")
  • Handle Length:
    5.6 cm (23/16")
  • L×W×H:
    19.1 x 5.4 x 4.9 cm (71/2 x 21/8 x 115/16")
  • Supplier No.:
    369062008

Specifications

About this item

Sterileware® scoops are the perfect shape and size for taking samples, especially from wide-mouth containers.

  • Suited for use with wide-mouth containers
  • Sterile
  • Polystyrene plastic construction
  • Provided in individually sealed polyethylene bag

Designed to hold various capacities, these one-piece sterile scoops are molded from high-impact polystyrene plastic, individually sealed and sterilized in a polyethylene bag, and ready for one-time use and disposal.

The 60 mL (2oz.) and 120 mL (4oz.) sampling scoops have a long handle that increases the distance between the hand and sample. When placed on a flat surface they remain upright and can be used as weighing boats. The bright red color reduces the chance of these scoops being accidentally left behind in a sample.

The distinctive color can also serve as a code to avoid cross contamination and provide a contrasting background to better view light colored samples. Made from FDA-grade materials, they are lot stamped for accurate lot tracing and shipped with a Certificate of Processing for Sterilization.

Customers Who Bought This Also Bought