Specifications
- Volume:1.2 cu. ft.
- Color:Clear
- Height:31.4 cm (12.4")
- Model:2.0
- Cat. no.:47750-984
- No. of Shelves:2
- Supplier No.:420721000
Specifications
About this item
The attractive design and sturdy construction of the Secador® line of desiccator cabinets assures easy and reliable storage of moisture- and light-sensitive products.
- Sturdy construction
- Easy and reliable storage
- Protects moisture- and UV-sensitive products
- Co-polyester construction
Choose from an extensive selection of sizes, styles and colors of desiccator cabinets to meet your specific dry storage needs
Large doors maximize access to interior space and have a patented seal design, tab latches and security loops for a padlock or tamper-evident seals. Perforated shelves facilitate circulation of dry air and a space for the desiccant is under the bottom shelf. An easy-to-view digital hygrometer is located in the door front.
1.0, 2.0, 3.0 and 4.0 vertical units have a compact 34.1W x 41.4 cmD footprint for space saving stacking up to 3 units high and a side swinging door that opens to the left. 4.0 horizontal units have a wider footprint, lower height and pull down “oven style” door. Replacement shelves are available and the 4.0 units can be converted from vertical to horizontal configuration or vice versa by turning the unit and ordering the appropriate number of shelves.
Durastar® co-polyester construction blocks 99% of UV light* and is resistant to staining, crazing and chemical attack. Amber color models reduce visible light penetration by over 50% making it the perfect choice for storing light sensitive materials like reagents, analytical standards and investigational compounds.