Order Entry
ContactUsLinkComponent
FILTER PAPER
Wet-Strengthened Qualitative Filter Paper, Grade 961, Ahlstrom
Catalog #: 47749-770
Supplier:  Ahlstrom
FILTER PAPER
Wet-Strengthened Qualitative Filter Paper, Grade 961, Ahlstrom
Catalog #: 47749-770
Supplier:  Ahlstrom

Specifications

  • Dimensions:
    12.5 cm dia.
  • Flow rate:
    Fast
  • Retention:
    19 µm
  • Load capacity:
    Very High
  • Cat. no.:
    47749-770
  • Supplier No.:
    9610-1250

Specifications

About this item

Ahlstrom 961 is a fast filtering grade with a creped surface and extremely high loading capacity.

It is recommended for the filtration of coarse and gelatinous precipitates.

Customers Who Bought This Also Bought