Order Entry
ContactUsLinkComponent
PRECISIONWARE BTL LDPE 16OZ
SP Bel-Art Precisionware® Leakproof Bottles, Narrow Mouth, LDPE, Bel-Art Products, a part of SP
Catalog #: 47748-082
PRECISIONWARE BTL LDPE 16OZ
SP Bel-Art Precisionware® Leakproof Bottles, Narrow Mouth, LDPE, Bel-Art Products, a part of SP
Catalog #: 47748-082

Specifications

  • Cap Size:
    28 mm
  • Capacity:
    500 ml (16.9 oz.)
  • Cat. no.:
    47748-082
  • Supplier No.:
    106219007

Specifications

About this item

Translucent, flexible bottles with leakproof design and matched polypropylene closures are FDA compliant and offer greater visibility than HDPE bottles.

  • Transparent, leakproof design
  • Flexible LDPE construction
  • Polypropylene closures
  • Greater visibility than HDPE bottles
  • Narrow mouth

Perfect for sampling and general storage of liquids, powders, or viscous fluids. Bottles 250 ml (8.4 oz.) and larger feature a recycling code molded into base.

Customers Who Bought This Also Bought