Order Entry
ContactUsLinkComponent
Disposable Respirator Storage Bags
Disposable Large Respirator Storage Bags, Allegro
Catalog #: 47747-250
Supplier:  Allegro
Small Business Enterprise
Disposable Large Respirator Storage Bags, Allegro
Catalog #: 47747-250
Supplier:  Allegro
Small Business Enterprise

Specifications

  • Ø:
    8"
  • W×L:
    11×21"
  • Cat. no.:
    47747-250
  • Supplier No.:
    4001-05

Specifications

About this item

Disposable storage bags with gussets help prevent cross contamination of any personal safety equipment such as respirators or ear muffs.

  • Pressure-sensitive seals for recording cleaning information
  • Single-Use

The large bags come with pressure-sensitive seals where cleaning information can be recorded.

Customers Who Bought This Also Bought