Order Entry
ContactUsLinkComponent
FILTER PAPER
Glass Microfiber Filter Paper, Grade 111, Ahlstrom
Catalog #: 47745-784
Supplier:  Ahlstrom
FILTER PAPER
Glass Microfiber Filter Paper, Grade 111, Ahlstrom
Catalog #: 47745-784
Supplier:  Ahlstrom

Specifications

  • Ø:
    11 cm
  • Flow rate:
    Fast
  • Retention:
    1.2 µm
  • Load capacity:
    Very High
  • Cat. no.:
    47745-784
  • Supplier No.:
    1110-1100

Specifications

About this item

Grade 111 is a binder-free microfiber glass filter paper that offers fine particle retention with high filtration speed and good loading capacity.

Customers Who Bought This Also Bought