Order Entry
ContactUsLinkComponent
BRAND* Semi-Micro Spectrophotometer Cuvettes
BRAND® Disposable Plastic Cuvettes, Acrylic and Polystyrene, BrandTech
Catalog #: 47744-642
Supplier:  Brandtech
BRAND* Semi-Micro Spectrophotometer Cuvettes
BRAND® Disposable Plastic Cuvettes, Acrylic and Polystyrene, BrandTech
Catalog #: 47744-642
Supplier:  Brandtech

Specifications

  • Description:
    Semi-Micro
  • Material:
    PS
  • Volume Range:
    1.5–3.0 mL
  • Supplier No.:
    759075D

Specifications

About this item

Disposable cuvettes are constructed of high-quality polymethyl methacrylate (PMMA or "acrylic") or polystyrene (PS), and are excellent for most spectrophotometry applications in the visible range

  • 10 mm disposable cuvettes with two optical sides
  • Fits most common spectrophotometers
  • Mold-cavity match for minimum variability

Acrylic cuvettes are suitable for assays between 300nm and 900nm. Choose polystyrene cuvettes for routine assays between 340nm and 900nm.

Cuvettes include an arrow mark to indicate direction of transmission and reduce variation. They are grouped by manufacturing mold cavity number to ensure the lowest variation in extinction coefficient, and are packaged in low dust, low scratch material.

They provide accurate, reliable results and are available in macro and semi-macro sizes to accommodate most sample volumes. Cuvettes are compatible with most spectrophotometers and photometers.

Ordering information: For disposible cuvettes for wavelengths below 300nm, see 47743-834 series.
Customers Who Bought This Also Bought