Specifications
- Description:Macro Cuvettes
- Cat. no.:47744-640
- Pathlength:10 mm
- Volume Range:2.5–4.5 mL
- Supplier No.:759170
Specifications
About this item
These disposable UV-transparent cuvettes fit most commercial 10mm-pathlength spectrophotometers and photometers. The cuvettes are manufactured from a proprietary plastic that is transparent from 220 - 900 nm, replacing quartz cuvettes for DNA, RNA, protein purity and quantification, and other UV/visible applications, while eliminating scratching, washing and sample cross-contamination.
- Replaces quartz for many common assays
- Enhanced chemical compatibility
- 70 µl minimum fill volume
- Available individually wrapped
These cuvettes also offer superior chemical compatibility for polar organic solvents, acids, and bases, including acetone, butanone, DMF, and concentrated hydrochloric acid. Cuvettes are mold-cavity matched for the lowest variation in extinction coefficient.
Ultra-micro cuvettes are available in two z-dimensions (8.5mm and 15mm) to match the beam height of most commercial spectrophotometers
The polypropylene cuvette rack features 16 numbered positions and can be autoclaved to 121 °C (250 °F).
A 70 µl minimum fill volume conserves sample. Round caps are available for ultra-micro cuvettes to provide a plug seal for short-term sample storage. Individually wrapped BIO-CERT* ultra-micro cuvettes are DNA-, DNase-, and RNase-free, providing a purity assurance not available with quartz cuvettes.