Accessories for Barnstead RO Systems, Thermo Scientific
Catalog #: 47742-614
Supplier: Thermo Fisher Scientific
Accessories for Barnstead RO Systems, Thermo Scientific
Catalog #: 47742-614
Supplier: Thermo Fisher Scientific
Specifications
- Description:Prefilter, 5 micron
- Cat. no.:47742-614
- Supplier No.:D502113
Specifications
About this item
Customers Who Bought This Also Bought
Methanol ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC, VWR Chemicals BDH®
Supplier: VWR International
$377.44 to $992.90
Acetone ≥99.5% (by GC, corrected for water content), OmniSolv®, Supelco®
Supplier: MilliporeSigma
$768.73 to $3,075.66
Buffers, Reference Standards, Color-Coded, Ricca Chemical Company
Supplier: Ricca Chemical
$160.67 to $1,597.95
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$635.20 to $2,541.50
n-Heptane ≥99% (n-heptane) [≥99.9% (saturated C7 hydrocarbons)], OmniSolv®, Supelco®
Supplier: MilliporeSigma
$971.75 to $3,463.97
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$684.63 to $2,738.51
Accessories for Barnstead GenPure xCAD Water Purification Systems, Thermo Scientific
Supplier: THERMO FISHER SCIENTIFIC MX
$2,009.11