Order Entry
ContactUsLinkComponent
LAMP, MERCURY
Short Arc Mercury Lamp Bulb, Bulbtronic©
Catalog #: 47736-696
Supplier:  Bulbtronics
LAMP, MERCURY
Short Arc Mercury Lamp Bulb, Bulbtronic©
Catalog #: 47736-696
Supplier:  Bulbtronics

Specifications

  • Description:
    Short Arc Mercury Lamp Bulb
  • Cat. no.:
    47736-696
  • Electrical:
    100 W
  • Bulb Life:
    300 h
  • Supplier No.:
    0000257

Specifications

About this item

Microscope replacement bulb.

Customers Who Bought This Also Bought