ContactUsLinkComponent
Flat Polyethylene Bags, 3 mil
Low Density Flat Poly Bags, Associated Bag
Catalog #: 47735-416
Supplier:  Associated Bag
Flat Polyethylene Bags, 3 mil
Low Density Flat Poly Bags, Associated Bag
Catalog #: 47735-416
Supplier:  Associated Bag
Specifications
  • Thickness:
    3 mil
  • Size:
    12×24"
  • Cat. no.:
    47735-416
  • Supplier No.:
    28-5-536
Specifications
About this item
Economical and reusable.
2 to 8 mil for light- to super-heavy weight uses. Not recommended for liquids.
Certifications: Meets FDA and USDA specifications for food contact.
Customers Who Bought This Also Bought