Order Entry
ContactUsLinkComponent
4oz Flip Top Vial
Flip-Top Containers with Lock Seal, Capitol Vial®
Catalog #: 46610-586
4oz Flip Top Vial
Flip-Top Containers with Lock Seal, Capitol Vial®
Catalog #: 46610-586

Specifications

  • Capacity:
    120 ml (4 oz.)
  • I.D.:
    43 mm (111/16")
  • Description:
    High Profile
  • Height:
    90 mm (31/2")
  • Supplier No.:
    04HPLS

Specifications

About this item

Triple-seal containers are sterile, airtight, and leakproof

Unique process physically closes containers in the mold during manufacture to ensure an airtight seal and zero particulates >0.5µ. Process also produces sterile containers that do not require a second means of sterilization for most applications. Color: clear.

Unique process physically closes containers in the mold during manufacture to ensure an airtight seal and zero particulates >0.5µ. Process also produces sterile containers that do not require a second means of sterilization for most applications. Color: clear.

Nonabsorptive, nonaerosoling containers are made of FDA-approved, food grade 100% polypropylene. Hinged top enables opening and closing with one hand. Ideal for storing and packaging a wide range of solids and liquids in applications such as medical device packaging, reagent packaging, solid sample storage, and water testing. Containers feature a lock seal tab that allows the user to lock the lid to the base of the container.

Certifications: Containers conform to FDA and SAMHSA specifications.
Customers Who Bought This Also Bought