PYREX® Volumetric Pipettes, Class A, Serialized, Corning
Catalog #: 46610-102
Supplier: Corning
Product available on GSA Advantage®
Specifications
- Capacity:1 ml
- Description:PYREX® volumetric pipettes, class A, serialized
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:I-RECs, ISO 14001 - Monterrey
- Cat. no.:46610-102
- Supplier No.:7103C-1
Specifications
About this item
These PYREX® volumetric pipettes are calibrated to Class A tolerances in accordance with ASTM E-542 and ASTM E-969.
- Reusable precision pipettes
- Color-coded markings are enameled onto the glass
PYREX® serialized/certified volumetric glassware is now tested and calibrated in an ISO/IEC 17025 certified laboratory.
Each pipette is individually serialized and supplied with a Certificate of Identification and capacity, traceable to NIST standards. These precision PYREX® volumetric pipettes are manufactured and calibrated 'To Contain' (TC: bottom line) and 'To Deliver' (TD: top line) to specifications for Class A volumetric ware.
Customers Who Bought This Also Bought