VWR® Advanced SPP Bouffant Caps
Catalog #: 414005-000
Supplier: VWR International
Specifications
- Ø:53 cm (21")
- Color:White
- Description:Latex-free
- Particle shedding (Helmke Drum):Level I
- Weight:14.0 g/m2
Specifications
About this item
Bouffant caps are manufactured from strong, breathable, spunbonded polypropylene (SPP) fabric.
- First-line barrier
- Higher basis weight for increased protection
- Non-linting
They protect against dirt, grime, and certain dry particulates in nonhazardous environments
Bouffant caps are manufactured and rigorously tested in an ISO Certified facility under stringent process controls to ensure that each garment meets exacting quality standards. Products are validated through independent lab testing.
Caps are packaged 100 per pack, 10 packs per case.
Customers Who Bought This Also Bought