VWR® Irradiated Beard Cover
Catalog #: 414004-506
Supplier: VWR International
Specifications
- Clothing Size:One size
- Ø:45.7 cm (18")
- Sterile:+
- Color:White
- Tearing strength:1.6 (Widthwise Yarns)
- Tensile strength:5.9 (Filling)
- Particle shedding (Helmke Drum):Level I
- Weight:16.82 g
Specifications
About this item
Beard cover is manufactured from strong, lightweight, and breathable spunbonded polypropylene (SPP) fabric, which protects against dirt, grime, and certain dry particulates in nonhazardous environments.
- Strong, lightweight, breathable spunbonded polypropylene
- Lowest level of particle shedding, non-linting
- First-line barrier
- Gamma irradiated to SAL of 10-4
To address the concern of bio-burden contamination, the beard cover is gamma irradiated to a Sterility Assurance Level (SAL) of 10-4. Manufactured and packaged in an ISO Class 8 (FED-STD-209E Class 100,000/M3.5) cleanroom.
Certifications: Beard cover is tested and manufactured in an ISO Certified facility under stringent process controls to ensure that each product meets exacting quality standards and performs to specification. Products are validated through independent lab testing.
Ordering information: Packaged in sealed polyethylene bags, 10 per bag, contained within a larger carton liner. Each case is supplied with a certificate of irradiation.
Customers Who Bought This Also Bought