DRIVE UNIT FOR MANDRELL 220V
Catalog #: 37000-310
Supplier: Koehler
Customers Who Bought This Also Bought
Formalin 10%, VWR®, Reagent Grade for histology, neutral buffered
Supplier: VWR International
$109.33 to $364.87
Ethanol denatured, anhydrous (denatured with methanol and ethyl acetate)
Supplier: INNOVA CHEMICALS CA
$468.00