EXISTENT GUM EVAP BTH 3 UNT220
Catalog #: 37000-240
Supplier: Koehler
Customers Who Bought This Also Bought
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
635,20 $ to 2 541,50 $
Methanol ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC, VWR Chemicals BDH®
Supplier: VWR International
377,44 $ to 992,90 $
Sulfuric acid 2.0 N in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
119,71 $ to 1 363,18 $
Pentane ≥98.0%, Laboratory Reagent, VWR Chemicals BDH®
Supplier: VWR International
500,19 $ to 1 790,36 $