VENTED CORK
Catalog #: 37000-174
Supplier: Koehler
Customers Who Bought This Also Bought
Glycerine ≥99.7%, Laboratory Reagent, VWR Chemicals BDH®
Supplier: VWR International
881,80 $ to 2 807,56 $
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
684,63 $ to 2 738,51 $
Plastic Forceps, with Jaw Grips, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
79,44 $ to 83,03 $
VeriSpec™ Potassium - K⁺ Standard for Ion Chromatography 1000 ppm in H₂O, RICCA Chemical Company
Supplier: Ricca Chemical
170,97 $
Scalpel Handles; Feather®, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
54,46 $