SAYBOLT WAX CHROMOMETR 220/15
Catalog #: 37000-066
Supplier: Koehler
Customers Who Bought This Also Bought
Starch 1% (w/v) in aqueous solution for iodometric titrations mercury-free
Supplier: Ricca Chemical
100,00 $
Alcohol ≥70%, Reagent Grade (denatured with methanol and isopropanol), VWR Chemicals BDH®
Supplier: VWR International
131,08 $ to 405,43 $
Puritan® Standard Cotton Tipped Applicators, Wood Handle, Sterile, Puritan Medical Products
Supplier: Puritan Medical Products
68,23 $ to 471,36 $
Hydrochloric acid 36.5 - 38.0%, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
655,42 $ to 655,42 $