OXIDATION BATH 2 VESSEL 115V
Catalog #: 37000-022
Supplier: Koehler
Customers Who Bought This Also Bought
Glycerine ≥99.7%, Laboratory Reagent, VWR Chemicals BDH®
Supplier: VWR International
$881.80 to $2,807.56
Extran® MN 01 Powdered Detergent for Manual Washing, MilliporeSigma
Supplier: MilliporeSigma
$214.10 to $1,039.32
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$635.20 to $2,541.50
Methanol ≥99.8%, GR ACS, meets analytical specification for testing USP/NF monographs, Supelco®
Supplier: MilliporeSigma
$1,413.13
Scalpel Handles; Feather®, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
$54.46
1-Octanesulfonic acid sodium salt ≥99%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Supplier: VWR International
$814.96