Biohazard Warning Label, National Marker
Catalog #: 36434-434
Supplier: ACCUFORM MANUFACTURING, INC
Specifications
- Description:Biohazard warning label
- Material:Plastic
- Cat. no.:36434-434
- Dimensions:76.2×76.2 mm (3×3")
- Supplier No.:S52RL
Specifications
About this item
Displays biohazard symbol and word.
- Pressure-sensitive paper rolls can be written on to provide additional information
Ordering information: Each roll contains 500 individual labels.
Customers Who Bought This Also Bought