To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Color:Blue
- Description:pH 10.01 buffer
- Size:475 ml
- Supplier No.:910110
Specifications
About this item
Perform accurate and reproducible pH calibrations with Orion™ pH buffer bottles.
- All bottles feature pH/temperature tables printed on the labels for accurate calibrations and NIST traceability
- Manufactured under ISO 9000 quality standards
- Available in pint (475 ml) bottles or portable five packs of 2 oz. (5×60 ml) bottles
The pH 4.01, 5.00, 7.00, and 10.01 buffers are color-coded for ease of use. All Orion™ pH buffers are traceable to NIST standard reference materials.