Order Entry
ContactUsLinkComponent
pH Buffer Bulk Pack B
Orion™ Pure Water™ pH Buffers and pHISA™ Adjustor, Thermo Scientific
Catalog #: 34182-072
CAS Number:  
pH Buffer Bulk Pack B
Orion™ Pure Water™ pH Buffers and pHISA™ Adjustor, Thermo Scientific
Catalog #: 34182-072
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Buffer B, pH 4.10
  • Cat. no.:
    34182-072
  • Supplier No.:
    700402

Specifications

About this item

Sample well water, rain water, distilled/deionized water, boiler feed water, process water, and more with the Orion™ Pure Water™ pH Buffers and pHISA™ Adjustor

  • Increases measurement accuracy
  • Reduces electrode drift
  • Eliminates contamination of sample
  • For best results, a high quality pH electrode is recommended

It offers fast, accurate pH results in high purity and low ionic strength waters.

Ordering information: Kit contains special low ionic strength buffers for calibration and pHISA™ adjustor additive, to increase sample ionic strength without changing pH.
Customers Who Bought This Also Bought