ContactUsLinkComponent
Filling Solution
Orion™ pH Electrode Filling Solution, Thermo Scientific
Catalog #: 34108-020
CAS Number:  
Filling Solution
Orion™ pH Electrode Filling Solution, Thermo Scientific
Catalog #: 34108-020
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Fill Solution for Standard, Speciality, PerpHecT Ag/AgCl, Single Junction Refillable Green and Micro pH Electrodes, ORP Electrodes
  • Size:
    60 mL
  • Cat. no.:
    34108-020
  • Supplier No.:
    900011
Specifications
About this item
Use Orion™ pH Electrode Filling Solution with pH electrodes and reference electrodes.
Only use ROSS filling solution with ROSS™ and ROSS Ultra™ pH electrodes to prevent damage. When the single junction reference electrode or double junction reference electrode are used with Orion™ ion selective electrodes (ISE), refer to the recommended ISE filling solution listed in the ISE user guide.
Customers Who Bought This Also Bought