Order Entry
ContactUsLinkComponent
pH Electrode Cleaning Solution A
Orion™ pH Electrode Cleaning and Storage Solutions, Thermo Scientific
Catalog #: 34104-296
CAS Number:  
pH Electrode Cleaning Solution A
Orion™ pH Electrode Cleaning and Storage Solutions, Thermo Scientific
Catalog #: 34104-296
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Cleaning solution A for protein removal, with beaker and pipet
  • Size:
    4×30 ml bottles
  • Cat. no.:
    34104-296
  • Supplier No.:
    900021-WA

Specifications

About this item

Cleaning Solutions restore proper electrode performance and prolong the useful life of the electrode.

  • Restore electrode performance
  • Prolong usefulness
  • Unique formulations

Cleaning Solutions are identified by the letters A, B, C, and D. Each letter designates a unique formulation. Cleaning solution A is for removing protein contaminants. Cleaning solution B is for removing bacterial contaminants. Cleaning solution C is for general cleaning. Cleaning solution D is for removing oil and grease contaminants. Bottles include a small beaker to hold the solution and a plastic pipet to remove the old electrode filling solution.

Customers Who Bought This Also Bought