To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Capacity:45 ml (1.5 oz.)
- Dimensions:23 I.D. x 84 L mm (29/32 x 35/16")
- Color:Clear
- Cat. no.:33500-728
- Supplier No.:02CL
Specifications
About this item
Triple-seal containers are airtight, leakproof, nonabsorptive, and nonaerosoling.
Unique process physically closes vials in the mold during manufacture to ensure an airtight seal and zero particulates >0.5µm. Sterile.
Unique process physically closes vials in the mold during manufacture to ensure an airtight seal and zero particulates >0.5µm. Sterile.
Made of FDA-approved, food-grade 100% polypropylene. Hinged top permits opening and closing these flip-top vials with one hand. Ideal for storing and packaging a wide range of solids and liquids in applications such as medical device packaging, reagent packaging, solid sample storage, and water testing.