Qualatherm® 70G Cleanroom Gloves, 450 Wet/Dry Thermal Protection, QRP
Catalog #: 32885-780
Supplier: PIP
Specifications
- Ambidextrous/Hand-specific:Hand-specific
- Cleanroom class:100/ISO 5
- Glove Color:White
- Glove size:L
- Length:30.5 cm (12")
- Powdered/Powder-free:Powder-free
- Thickness:116 mil
- Cat. no.:32885-780
- Supplier No.:70GL
Specifications
About this item
The QRP 70G Qualatherm Cleanroom Mid-Temperature Gloves are RoHS compliant and have a PolyTuff silicon elastomer formulation that offers clean, liquid-proof mid-temperature protection for wet or dry environments.
- Packaged for ISO 5 (Class 100)
- Wet or dry contact temperatures from –78 to 450 °F (–61.4 to 232 °C)
- PolyTuff silicone elastomer is sulfur-free
- Excellent resistance to chemicals, solder, and fluxes
- Free from PCB, asbestos and fiberglass hazards
- Latex-free
- Non-particulating, cut and abrasion resistant
Seamless Comfort Curve™ paired Right Hand/Left Hand gloves minimize hand and finger fatigue. Micro-coated, high density, continuous filament nylon liner assures clean, easy donning and removal.
Applications of the 70G Qualatherm 450 °F mid-temperature cleanroom gloves include: diffusion furnaces, Quartz wafer boats, burn-in ovens, PWB handling after wave soldering, ceramics, porcelains, food processing and handling.
Customers Who Bought This Also Bought