Order Entry
ContactUsLinkComponent
Vacuum Gauge, Canner's Test
Canners Vacuum Guage, Jensen Instrument
Catalog #: 31757-128
Supplier:  Jensen Instrument
Canners Vacuum Guage, Jensen Instrument
Catalog #: 31757-128
Supplier:  Jensen Instrument

Specifications

  • Cat. no.:
    31757-128
  • Supplier No.:
    50462

Specifications

About this item

Canners Gauge is used for testing vacuum of packaged product such as canned food or drink.

  • Easy to use for destructive sampling tests
  • 1" graduations in Hg, inside 2" diameter case
  • Tip can be cleaned for multiple use, non-removable
  • The rubber gasket prevents air from entering the container during the test
  • Force the stainless steel needle through the top of the container while reading the gauge
Customers Who Bought This Also Bought