WaterPro® PS Polishing System, Labconco
Catalog #: 30181-416
Supplier: Labconco
Product available on GSA Advantage®
Specifications
- Description:Polishing Kit (Required)
- Cat. no.:30181-416
- Supplier No.:9047101
Specifications
About this item
WaterPro PS Polishing Stations deliver Type I, 18.2 megohm-cm water from the dispensing valve or optional dispensing gun (refer to descriptions for models that include gun) at a typical rate of 1.8 liters per minute depending on feedwater flow rate and pressure, and installed purification modules.
The control panel includes a timed dispense feature that allows unattended dispensing of pure water without risk of overfilling. The dispenser shuts off automatically once user-set time has elapsed.
Ordering information: Polishing kit (30181-416) is also required for operation and is sold separately. For available accessories, contact your Avantor representative.
Customers Who Bought This Also Bought