Corning® 96-Well Polypropylene Storage Blocks
Catalog #: 29445-164
Supplier: Corning
Product available on GSA Advantage®
Specifications
- Sterility:Sterile
- Well Volume:2 mL
- Color:Clear
- Description:Storage block, 96-well, natural, square well
- Cat. no.:29445-164
- Well Shape:V-Bottom
- Supplier No.:3960
Specifications
About this item
Plates are available in V-bottom and round-bottom formats to meet a wide variety of assay and storage needs.
Robolid combines the sealing ability of a storage mat with the rigidity of a plastic lid and is designed for repeated application and removal by automation and to prevent short-term evaporation. Robolid also features silicone sealing plugs for organic solvent resistance and low extractables.
All sterile products are certified RNase- and DNase-free. Built to the industry standard footprint.
Ordering information: Storage mats and sealing tapes sold separately.
Customers Who Bought This Also Bought