Corning® 96-Well Plates, Polypropylene, Without Lids
Catalog #: 29444-104
Supplier: Corning
Product available on GSA Advantage®
Specifications
- Sterility:Non sterile
- Well Volume:360 µl
- Color:Clear
- Description:Storage plate, 96-well, natural, round well
- Well Shape:Round-Bottom
- Supplier No.:3365
Specifications
About this item
Corning polypropylene microplates offer both small volume and large volume (blocks) well formats to meet assay and storage requirements.
- Flat, round, or V-shaped well bottom
- Features uniform skirt heights for greater robotic gripping surface
- Solvent resistant polypropylene provides compatibility with many common organic solvents (e.g., DMSO, ethanol, methanol)
- RNase-/DNase-free
- Available sterile or non sterile
Ordering information: Plates are packaged 25 per pack, 4 packs per case. Nonsterile 450 µl plate is packaged 10 plates per bag and 5 bags per case.
Customers Who Bought This Also Bought