Order Entry
ContactUsLinkComponent
Costar* Brand 96-Well Solid Plates
Corning® 96-Well Solid Black and White Polystyrene Microplates, Corning
Catalog #: 29444-041
Supplier:  Corning
   Low Manufacturing Impact
Product available on GSA Advantage®
Costar* Brand 96-Well Solid Plates
Corning® 96-Well Solid Black and White Polystyrene Microplates, Corning
Catalog #: 29444-041
Supplier:  Corning
   Low Manufacturing Impact
Product available on GSA Advantage®

Specifications

  • Sterility:
    Sterile
  • Well Volume:
    360 µl
  • Color:
    White
  • Description:
    Tissue culture treated, flat bottom, with lid
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Material:
    PS
  • No. of wells:
    96
  • Environmental certification:
    RECs, ISO 14001 - Kennebunk
  • Cat. no.:
    29444-041
  • Packaging:
    20/bag
  • Supplier No.:
    3917

Specifications

About this item

These 96-well solid black and white polystyrene microplates are designed to reduce well-to-well cross-talk.

White microplates enhance luminescent signals and have low background luminescence.

Black microplates have low background fluorescence and minimize light scattering.

Customers Who Bought This Also Bought