Cell Culture Storage Bottles, Polystyrene, Corning
Catalog #: 29442-878
Supplier: Corning
Product available on GSA AdvantageĀ®
Specifications
- Capacity:500 ml (16.9 oz.)
- Cat. no.:29442-878
- Supplier No.:8393
Specifications
About this item
Disposable polystyrene bottles for storage of media, buffers and other aqueous solutions.
- Low profile, easy grip style has sides that facilitate handling
- Plug seal caps (45 mm) provide an airtight seal and help minimise the risk of contamination
- Bottles can be used with CorningĀ® vacuum filter systems
- Sterile, certified non-pyrogenic
Customers Who Bought This Also Bought